Learn More
Avanti Polar Lipids 18:1 DAP OIL 1,2-DIOLE 200MG
Catalog No. NC1978650
Encompass
18:1 DAP {OIL} 1,2-dioleoyl-3-dimethylammonium-propane (DODAP), >99% 200mg CAS#127512-29-2 C41H77NO4
Catalog No. NC1978650 Supplier Avanti Polar Lipids Supplier No. 890850O200MG
18:1 DAP {OIL} 1,2-dioleoyl-3-dimethylammonium-propane (DODAP), >99% 200mg CAS#127512-29-2 C41H77NO4