Learn More
Avanti Polar Lipids 18:1 LYSO PA (CHCL3) 25MG
Supplier:  Avanti Polar Lipids 857130C25MG
Encompass
18: 1 Lyso PA, 1-Oleoyl-2-Hydroxy-sn-Glycero-3-Phosphate (Sodium Salt), 25 mg, Molecular Weight: 458.51, Molecular Formula: C21H40O7PNa, Percent Composition: C 55.01% H 8.79% Na 5.01% O 24.43% P 6.76%
Catalog No. NC9860653