Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avanti Polar Lipids 18:1 Lyso PG, 500 mg, Liquid, 25 mg/mL, Chloroform, 5 x 4 mL ampule, -20°C, CAS 326495-24-3

Catalog No. NC1662009
Encompass

18:1 Lyso PG is a high-purity, synthetic lysophospholipid detergent provided by Avanti Polar Lipids. This nonbilayer-forming lipid is crucial in membrane research due to its detergent-like properties, effectively disrupting membrane integrity. It plays a pivotal role as a substrate for lysophosphatidylglycerol acyltransferase 1 (LPGAT1) in phosphoglycerol synthesis and is frequently used as a lipid standard in liquid chromatography-mass spectrometry (LC-MS) method optimization.

  • High purity greater than 99%
  • Liquid physical form
  • Utilizes chloroform as solvent
  • Packaged in 5 x 4 mL ampules
  • Unit size of 500 mg
  • Concentration of 25 mg/mL
  • Requires shipping on dry ice
  • Stored at -20°C
  • Empirical formula: C24H46O9PNa
  • CAS number: 326495-24-3
  • Molecular weight of 532.58 g/mol
  • MDL number: MFCD00672604

Catalog No. NC1662009 Supplier Avanti Polar Lipids Supplier No. 858125C500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

18:1 Lyso PG is a high-purity, synthetic lysophospholipid detergent provided by Avanti Polar Lipids. This nonbilayer-forming lipid is crucial in membrane research due to its detergent-like properties, effectively disrupting membrane integrity. It plays a pivotal role as a substrate for lysophosphatidylglycerol acyltransferase 1 (LPGAT1) in phosphoglycerol synthesis and is frequently used as a lipid standard in liquid chromatography-mass spectrometry (LC-MS) method optimization.

  • High purity greater than 99%
  • Liquid physical form
  • Utilizes chloroform as solvent
  • Packaged in 5 x 4 mL ampules
  • Unit size of 500 mg
  • Concentration of 25 mg/mL
  • Requires shipping on dry ice
  • Stored at -20°C
  • Empirical formula: C24H46O9PNa
  • CAS number: 326495-24-3
  • Molecular weight of 532.58 g/mol
  • MDL number: MFCD00672604

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.