Learn More
Avanti Polar Lipids 18:1 Lyso PG, 326495-24-3, MFCD00674367, powder, 500mg
Catalog No. NC1599349
Encompass
Molecular Formula C24H46O9PNa, Purity >99% LPG; may contain up to 10% of the 2-LPG isomer, Formula Weight 532.580, Storage Temperature -20°C, 1-oleoyl-2-hydroxy-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt)
Catalog No. NC1599349 Supplier Avanti Polar Lipids Supplier No. 858125P500MG
Molecular Formula C24H46O9PNa, Purity >99% LPG; may contain up to 10% of the 2-LPG isomer, Formula Weight 532.580, Storage Temperature -20°C, 1-oleoyl-2-hydroxy-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt)