Learn More
Avanti Polar Lipids 1-oleoyl-2-hydroxy-sn-glycero-3-phospho-L-serine (sodium salt), 326589-90-6, MFCD00674368, 5mg
Catalog No. NC1336956
Encompass
Molecular Formula: C24H45NO9PNa, Formula Weight: 545.579, Purity >99% LPS; may contain up to 10% of the 2-LPS isomer.
Catalog No. NC1336956 Supplier Avanti Polar Lipids Supplier No. 858143P5MG
Molecular Formula: C24H45NO9PNa, Formula Weight: 545.579, Purity >99% LPS; may contain up to 10% of the 2-LPS isomer.