Learn More
Avanti Polar Lipids 18:1 NBD PE, 384832-99-9, MFCD22495272, powder, 5mg
Supplier:  Avanti Polar Lipids 810145P5MG
Encompass
Molecular Formula C47H82N5O11P, Purity >99%, Formula Weight 924.155, Storage Temperature -20°C, 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt)
Catalog No. NC1544147