Learn More
Avanti Polar Lipids 18:1 TAP (DOTAP), Chloroform | 132172-61-3 | MFCD01310872 | 25 mg
Catalog No. NC0974782
Encompass
18:1 TAP (DOTAP), Chloroform | Purity: 99% | Mol Wt: 697.578 | 132172-61-3 | MFCD01310872 | 25 mg
Catalog No. NC0974782 Supplier Avanti Polar Lipids Supplier No. 890890C25MG
18:1 TAP (DOTAP), Chloroform | Purity: 99% | Mol Wt: 697.578 | 132172-61-3 | MFCD01310872 | 25 mg