Learn More
MSC Thomas & Betts B14-8F #8 Stud, 18 to 14 AWG Compatible, Noninsulated, Crimp Connection, Standard Fork Terminal 0.31" Fork Width, Blue, 3/4" Overall Length, 600 Volts, Copper Contacts
Catalog No. 501296824
Encompass
Thomas & Betts B14-8F #8 Stud, 18 to 14 AWG Compatible, Noninsulated, Crimp Connection, Standard Fork Terminal 0.31" Fork Width, Blue, 3/4" Overall Length, 600 Volts, Copper Contacts
Catalog No. 50-129-6824 Supplier MSC Supplier No. 54064548
Thomas & Betts B14-8F #8 Stud, 18 to 14 AWG Compatible, Noninsulated, Crimp Connection, Standard Fork Terminal 0.31" Fork Width, Blue, 3/4" Overall Length, 600 Volts, Copper Contacts