Learn More
Chemglass Life Sciences Screw Cap, Open Top, 18-400, Black, with PTFE Faced Silicone Septa
SDP
Catalog No. CG491003
Encompass_Preferred
Screw thread size: 18-400, BLACK. Open top polypropylene screw caps supplied with PTFE faced silicone septa. Supplied in packages of 144.
Catalog No. CG491003 Supplier Chemglass Life Sciences Supplier No. CG491003
Screw thread size: 18-400, BLACK. Open top polypropylene screw caps supplied with PTFE faced silicone septa. Supplied in packages of 144.