Learn More
Chemglass Life Sciences 18-415 G.P.I. Glass Thread, 19mm OD Tubing, 125mm OAL
SDP
Catalog No. CG19005
Encompass_Preferred
18-415 G.P.I. Glass Thread, 19mm OD Tubing, 125mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread. PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.
Catalog No. CG19005 Supplier Chemglass Life Sciences Supplier No. CG19005
18-415 G.P.I. Glass Thread, 19mm OD Tubing, 125mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread. PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.