Learn More
Laboratory Equipment Co PHCbi MDF-DU502VH-PA Ultra-Low Freezer, 18.6 cu. ft. Capacity, -50 to -86 °C Temperature Range, 220V
SDP
Catalog No. NC3442618
Encompass_Preferred
The PHCbi MDF-DU502VH-PA VIP ECO Ultra-Low Temperature Freezer features energy-efficient storage for biological samples at -50 to -86 °C. It utilizes natural refrigerants and an advanced frost control system, ensuring stable temperatures while minimizing environmental impact. With a capacity of 18.6 cu ft and a user-friendly design, it is perfect for biorepositories and medical research facilities aiming for reliable cold chain storage.
Catalog No. NC3442618 Supplier Laboratory Equipment Co Supplier No. MDFDU502VHPA
Add to Cart
Add to Cart
Request a Quote
The PHCbi MDF-DU502VH-PA VIP ECO Ultra-Low Temperature Freezer features energy-efficient storage for biological samples at -50 to -86 °C. It utilizes natural refrigerants and an advanced frost control system, ensuring stable temperatures while minimizing environmental impact. With a capacity of 18.6 cu ft and a user-friendly design, it is perfect for biorepositories and medical research facilities aiming for reliable cold chain storage.