Learn More
Ace Glass, Inc. 18/7 BALL JOINT (GLASSBLOWER)
SDP
Catalog No. 50975268
Encompass_Preferred
Full length, standard wall 18/7 ball joint. This ACE ball joint is precision ground and vacuum leak-tested for fit and interchangeability.
Catalog No. 50-975-268 Supplier Ace Glass, Inc. Supplier No. 765516
Full length, standard wall 18/7 ball joint. This ACE ball joint is precision ground and vacuum leak-tested for fit and interchangeability.