Learn More
Chemglass Life Sciences 18/7 Ball Joint, Clamp Size: 18, Tubing OD: 9.80-10.20mm
SDP
Catalog No. CG12005
Encompass_Preferred
18/7 Ball Joint, Clamp Size: 18, Tubing OD: 9.80-10.20mm.Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12005 Supplier Chemglass Life Sciences Supplier No. CG12005
18/7 Ball Joint, Clamp Size: 18, Tubing OD: 9.80-10.20mm.Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.