Learn More
Chemglass Life Sciences Joint, 18/7, Socket, Clamp Size 18, 9.80-10.20mm Tubing
SDP
Catalog No. CG12105
Encompass_Preferred
Joint, 18/7, Socket, Clamp Size 18, 9.80-10.20mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12105 Supplier Chemglass Life Sciences Supplier No. CG12105
Joint, 18/7, Socket, Clamp Size 18, 9.80-10.20mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.