Learn More
McMaster-Carr 18-8 SS SOCKET 16MM LONG
Supplier:  McMaster-Carr 91292A135
Encompass
The McMaster-Carr 91292A135 is a stainless steel hex drive flat head screw with a thread diameter of 1/4"-20. It has a head height of 3/16" and a nominal length of 1-1/2". The screw material is made of 18-8 stainless steel for corrosion resistance. The drive size is 5/16" hex and it can be used with a hex wrench or hex driver. The flat head provides a low profile while still allowing a tool to drive it flush. This screw meets ASME B18.3 standards and has a tensile strength of 85,000 psi. It can be used for various securing and fastening applications.
Catalog No. NC1536421