Learn More
Chemglass Life Sciences 18/9 Ball Joint, Clamp Size: 18, Tubing OD: 12.50-12.90mm
SDP
Catalog No. CG12006
Encompass_Preferred
18/9 Ball Joint, Clamp Size: 18, Tubing OD: 12.50-12.90mm. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12006 Supplier Chemglass Life Sciences Supplier No. CG12006
18/9 Ball Joint, Clamp Size: 18, Tubing OD: 12.50-12.90mm. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.