Learn More
Ace Glass, Inc. 18/9 ball joint, stainless steel, 1/4" male NPT
SDP
Catalog No. 50975305
Encompass
18/9 ball joint, fabricated from 18-8 type 303 stainless steel, is fully interchangeable with ACE glass joints, and has same size I.D. Shank is machined to 1/4 male NPT.
Catalog No. 50-975-305 Supplier Ace Glass, Inc. Supplier No. 7658220
18/9 ball joint, fabricated from 18-8 type 303 stainless steel, is fully interchangeable with ACE glass joints, and has same size I.D. Shank is machined to 1/4 male NPT.