Learn More
Chemglass Life Sciences 18/9 O-Ring Ball Joint, Clamp Size: 18, O-Ring Size: 112, Tubing OD: 12.50-12.90mm
SDP
Catalog No. CG12205
Encompass_Preferred
18/9 O-Ring Ball Joint, Clamp Size: 18, O-Ring Size: 112,Tubing OD: 12.50-12.90mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.
Catalog No. CG12205 Supplier Chemglass Life Sciences Supplier No. CG12205
18/9 O-Ring Ball Joint, Clamp Size: 18, O-Ring Size: 112,Tubing OD: 12.50-12.90mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.