Learn More
Chemglass Life Sciences Joint, 18/9, Polished Socket, Clamp Size 18, 12.50-12.90mm Tubing
SDP
Catalog No. CG121P02
Encompass_Preferred
Joint, 18/9, Polished Socket, Clamp Size 18, 12.50-12.90mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.
Catalog No. CG121P02 Supplier Chemglass Life Sciences Supplier No. CG121P02
Joint, 18/9, Polished Socket, Clamp Size 18, 12.50-12.90mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.