Learn More
Chemglass Life Sciences 18/9 Socket Joint, 13.0mm OD Tube, Clamp Size = 18
SDP
Catalog No. 501531912
Encompass_Preferred
18/9 Socket Joint, 13.0mm OD Tube, Clamp Size = 18. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. 50-153-1912 Supplier Chemglass Life Sciences Supplier No. CGB12106
18/9 Socket Joint, 13.0mm OD Tube, Clamp Size = 18. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.