Learn More
Chemglass Life Sciences Joint, 18/9, Socket, Clamp Size 18, 12.50-12.90mm Tubing
SDP
Catalog No. CG12106
Encompass_Preferred
Joint, 18/9, Socket, Clamp Size 18, 12.50-12.90mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12106 Supplier Chemglass Life Sciences Supplier No. CG12106
Joint, 18/9, Socket, Clamp Size 18, 12.50-12.90mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.