Learn More
Ace Glass, Inc. 18/9 socket joint, stainless steel, 1/4" male NPT
SDP
Supplier:  Ace Glass, Inc. 7658222
Encompass
18/9 socket joint, fabricated from 18-8 type 303 stainless steel, is fully interchangeable with ACE glass joints, and has same size I.D. Shank is machined to male NPT.
Catalog No. 50-979-423