Learn More
Ace Glass, Inc. 18/9 SS PINCH CLAMP 18/9 SS PINCH CLAMP
SDP
Catalog No. 50979429
Encompass
Stainless Steel screwlock pinch clamp for use with 18/9 o-ring or ball & socket joints.
Catalog No. 50-979-429 Supplier Ace Glass, Inc. Supplier No. 766910
Stainless Steel screwlock pinch clamp for use with 18/9 o-ring or ball & socket joints.