Learn More
Medchemexpress LLC 18-Azido-stearic acid | 1529763-58-3 | C18H35N3O2 | 100 MG
SDP
Catalog No. 5000372941
Encompass_Preferred
18-Azido-stearic acid is a click chemistry reagent featuring an azide group. It functions as a hydrophobic bioconjugation linker, modifiable at the azido-position using Click-chemistry. This reagent can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN group-containing molecules.
  • Click chemistry reagent with an azide group
  • Functions as a hydrophobic bioconjugation linker
  • Can be modified at the azido-position via Click-chemistry
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-72941 Supplier Medchemexpress LLC Supplier No. HY151692100MG
18-Azido-stearic acid is a click chemistry reagent featuring an azide group. It functions as a hydrophobic bioconjugation linker, modifiable at the azido-position using Click-chemistry. This reagent can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN group-containing molecules.
  • Click chemistry reagent with an azide group
  • Functions as a hydrophobic bioconjugation linker
  • Can be modified at the azido-position via Click-chemistry
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC)