Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 18-Azido-stearic acid | 1529763-58-3 | C18H35N3O2 | 25 MG
SDP

Catalog No. 5000374551
Encompass_Preferred

18-Azido-stearic acid is a click chemistry reagent containing an azide group. It can be used as a hydrophobic bioconjugation linker and further modified at the azido-position using Click-chemistry. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups or ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.

  • Purity: 95.0%
  • Appearance: Solid
  • Color: White to off-white
  • Solubility (in vitro): DMSO: 100 mg/mL (307.23 mM; need ultrasonic)

Catalog No. 50-003-74551 Supplier Medchemexpress LLC Supplier No. 25MGHY151692
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

18-Azido-stearic acid is a click chemistry reagent containing an azide group. It can be used as a hydrophobic bioconjugation linker and further modified at the azido-position using Click-chemistry. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups or ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.

  • Purity: 95.0%
  • Appearance: Solid
  • Color: White to off-white
  • Solubility (in vitro): DMSO: 100 mg/mL (307.23 mM; need ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.