Learn More
Medchemexpress LLC 18-Azido-stearic acid | 1529763-58-3 | C18H35N3O2 | 5 MG
SDP
Catalog No. 5000372171
Encompass_Preferred
18-Azido-stearic acid is a versatile click chemistry reagent featuring an azide group. It functions as a hydrophobic bioconjugation linker, particularly when utilized with N-Myristoyltransferase, allowing for further modifications via Click-chemistry. It effectively participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Contains an azide group for click chemistry applications
  • Functions as a hydrophobic bioconjugation linker
  • Can be further modified via click-chemistry
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-72171 Supplier Medchemexpress LLC Supplier No. HY1516925MG
18-Azido-stearic acid is a versatile click chemistry reagent featuring an azide group. It functions as a hydrophobic bioconjugation linker, particularly when utilized with N-Myristoyltransferase, allowing for further modifications via Click-chemistry. It effectively participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Contains an azide group for click chemistry applications
  • Functions as a hydrophobic bioconjugation linker
  • Can be further modified via click-chemistry
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC)