Learn More
Medchemexpress LLC 18-azido-stearic acid | 1529763-58-3 | MFCD35066654 | ≥95.0% | 325.49 g/mol | C18H35N3O2 | 10 MG
SDP
Catalog No. 5000285937
Encompass_Preferred
18-Azido-stearic acid is an azide-functionalized long-chain fatty acid used as a hydrophobic bioconjugation linker for click chemistry. It enables efficient attachment of lipophilic tags or cargos to biomolecules via copper-catalyzed or strain-promoted azide-alkyne cycloaddition, and is supplied as a stable powder for laboratory research use.
  • Contains a terminal azide functional group for click reactions.
  • Compatible with CuAAC and SPAAC bioconjugation methods.
  • Hydrophobic stearic backbone facilitates membrane incorporation or lipophilic tagging.
  • Solid powder form with recommended storage for long-term stability.
  • Available in small milligram pack sizes suitable for research applications.
Catalog No. 50-002-85937 Supplier Medchemexpress LLC Supplier No. HY15169210MG
18-Azido-stearic acid is an azide-functionalized long-chain fatty acid used as a hydrophobic bioconjugation linker for click chemistry. It enables efficient attachment of lipophilic tags or cargos to biomolecules via copper-catalyzed or strain-promoted azide-alkyne cycloaddition, and is supplied as a stable powder for laboratory research use.
  • Contains a terminal azide functional group for click reactions.
  • Compatible with CuAAC and SPAAC bioconjugation methods.
  • Hydrophobic stearic backbone facilitates membrane incorporation or lipophilic tagging.
  • Solid powder form with recommended storage for long-term stability.
  • Available in small milligram pack sizes suitable for research applications.