Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 18-Crown-6

Catalog No. 503081489 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1489 100 g
28-125-5G 5 g
28-125-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-308-1489 Supplier Sigma Aldrich Supplier No. 28125100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

18-Crown-6

Specifications

CAS 17455-13-9
Melting Point 38°C to 41°C; 42°C to 45°C
Molecular Formula C12H24O6
Linear Formula C12H24O6
MDL Number MFCD00005113
Quantity 100 g
Synonym 18C6; 1,4,7,10,13,16-Hexaoxacyclooctadecane
Molecular Weight (g/mol) 264.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.