Learn More
AdipoGen 18-Crownether-6
SDP
Supplier:  AdipoGen CDXC0143G010
Encompass_Preferred
Chemical. CAS 17455-13-9. Formula C12H24O6. Molecular Weight 264.32. 18-Crownether-6 18-crown-6 is ionophoric and used as an efficient phase transfer catalyst and as a complexing agent with a variety of small cations. It is involved in the synthesis of diaryl ethers, diaryl thioethers and diarylamines mediated by potassium fluoride-alumina and 18-crown-6. Used to solubilizing simple metal salts in nonpolar and dipolar aprotic solvents where solvation of the anionic portion of the salt should be minimal. It facilitates the solubility of potassium permanganate in benzene, which is used for oxidizing the organic compounds. It is used to accelerate various substitution reactions as well as enhances the power of nucleophiles such as potassium acetate. It is utilized in the alkylation reactions in the presence of potassium carbonate, N-alkylation of glutarimide and succinimide with dimethylcarbonate.
Catalog No. 50-256-1458