Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ 18% Ethanol
GREENER_CHOICE

Catalog No. A65777BE
Change view
Click to view available options
Quantity:
5 L
20 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A65777BE 200 L
A65777BA 5 L
A65777BB 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A65777BE Supplier Gibco™ Supplier No. A65777BE
Add to Cart
Add to Cart

Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.

Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.

Features

  • Available in a range of pack sizes
  • Established shelf life; set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: Aegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Raw Material: 200 Proof Ethanol
CAS 64-17-5
Quantity 200 L
Packaging Bioprocess Container in Secondary Packaging
Boiling Point Information not available.
Molecular Weight (g/mol) Raw Material: 46.07
Percent Purity Not Available
Grade ACS/BP/EP/JP/USP

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.