Learn More
MSC Southwire 411010508 18 AWG, 16 Strand, Black Machine Tool Wire PVC, Acid, Moisture and Oil Resistant, 500 Ft. Long
Catalog No. 501296742
Encompass
Southwire 411010508 18 AWG, 16 Strand, Black Machine Tool Wire PVC, Acid, Moisture and Oil Resistant, 500 Ft. Long
Catalog No. 50-129-6742 Supplier MSC Supplier No. 54092697
Southwire 411010508 18 AWG, 16 Strand, Black Machine Tool Wire PVC, Acid, Moisture and Oil Resistant, 500 Ft. Long