Learn More
Apexbio Technology LLC 18α-Glycyrrhetinic acid 1449-05-4 10mg
SDP
Catalog No. 5000093572
Encompass_Preferred
18 -Glycyrrhetinic acid (CAS 1449-05-4) is a dietary-derived small molecule known to inhibit NF- B signaling while promoting proteasome activation Experimental studies have demonstrated that it enhances lifespan and exhibits anti-aggregation properties in multicellular organisms The compound also induces apoptosis in cellular models These activities highlight its potential utility in pharmacological research related to lifespan extension protein aggregation disorders and apoptosis-regulated diseases
Catalog No. 50-000-93572 Supplier Apexbio Technology LLC Supplier No. A474910
18 -Glycyrrhetinic acid (CAS 1449-05-4) is a dietary-derived small molecule known to inhibit NF- B signaling while promoting proteasome activation Experimental studies have demonstrated that it enhances lifespan and exhibits anti-aggregation properties in multicellular organisms The compound also induces apoptosis in cellular models These activities highlight its potential utility in pharmacological research related to lifespan extension protein aggregation disorders and apoptosis-regulated diseases