Learn More
Cayman Chemical 18β-Glycyrrhetinic Acid, Purity: ≥98%, CAS: 471-53-4, 1 g
SDP
Catalog No. 502952028
Encompass_Preferred
A major metabolite of glycyrrhizin, a constituent of licorice that exhibits anti-ulcerative, anti-inflammatory, and immunomodulatory properties; at 100 mg/kg/day, reduces lipid peroxidation and increases antioxidant activity in diabetic rats; suppresses LPS-induced TNF-α production and NF-κB activation in mouse macrophages 100-200 μM
Catalog No. 50-295-2028 Supplier Cayman Chemical Supplier No. 118451 G
A major metabolite of glycyrrhizin, a constituent of licorice that exhibits anti-ulcerative, anti-inflammatory, and immunomodulatory properties; at 100 mg/kg/day, reduces lipid peroxidation and increases antioxidant activity in diabetic rats; suppresses LPS-induced TNF-α production and NF-κB activation in mouse macrophages 100-200 μM