Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

18-β-Glycyrrhetinic Acid, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
471-53-4
C30H46O4
470.69
MFCD00003706
MPDGHEJMBKOTSU-YKLVYJNSSA-N
enoxolone,glycyrrhetinic acid,uralenic acid,glycyrrhetic acid,18-beta-glycyrrhetinic acid,glycyrrhetin,arthrodont,jintan,18beta-glycyrrhetinic acid,rhetinic acid
10114
CHEBI:30853
(2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylic acid
CC1(C2CCC3(C(C2(CCC1O)C)C(=O)C=C4C3(CCC5(C4CC(CC5)(C)C(=O)O)C)C)C)C
471-53-4
C30H46O4
470.69
MFCD00003706
MPDGHEJMBKOTSU-YKLVYJNSSA-N
enoxolone,glycyrrhetinic acid,uralenic acid,glycyrrhetic acid,18-beta-glycyrrhetinic acid,glycyrrhetin,arthrodont,jintan,18beta-glycyrrhetinic acid,rhetinic acid
10114
CHEBI:30853
(2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylic acid
CC1(C2CCC3(C(C2(CCC1O)C)C(=O)C=C4C3(CCC5(C4CC(CC5)(C)C(=O)O)C)C)C)C
Melting Point 292°C to 295°C
Specific Rotation + 165
Specific Rotation Condition +165° (20°C c=1,CHC3,on dry ba)
Loss on Drying 1% max. (105°C, 4 hrs)
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color White
Sulfated Ash 0.2% max.
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Merck Index 15, 3644
Formula Weight 470.69
Physical Form Crystalline Powder
Percent Purity ≥98.0% (on anhydrous substance)
Chemical Name or Material 18-β-Glycyrrhetinic acid, 98+%
Show More Show Less

EINECSNumber : 207-444-6

RTECSNumber : RK0180000

TSCA : TSCA