Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

18-beta-Glycyrrhetinic acid, 98+%

Catalog No. p-139162
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120150050 5 g Glass bottle
AC120150250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC120150050 Supplier Thermo Scientific Chemicals Supplier No. 120150050
Only null left
Add to Cart

CAS: 471-53-4 | C30H46O4 | 470.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-53-4
Molecular Formula C30H46O4
Molecular Weight (g/mol) 470.69
MDL Number MFCD00003706,MFCD00066716
InChI Key MPDGHEJMBKOTSU-YKLVYJNSSA-N
Synonym enoxolone, glycyrrhetinic acid, uralenic acid, glycyrrhetic acid, 18-beta-glycyrrhetinic acid, glycyrrhetin, arthrodont, jintan, 18beta-glycyrrhetinic acid, rhetinic acid
PubChem CID 10114
ChEBI CHEBI:30853
IUPAC Name (2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylic acid
SMILES CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 292°C to 295°C
Color White
Loss on Drying 1% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 3644
Specific Rotation Condition +165° (20°C c=1,CHC3,on dry ba)
Specific Rotation + 165
Formula Weight 470.69
Percent Purity 98+%
Related Substances 2% max. (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 18-β-Glycyrrhetinic acid, 98+%
Show More Show Less
EINECSNumber 207-444-6
RTECSNumber RK0180000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.