Learn More
Medchemexpress LLC 18β-Glycyrrhetyl-3-O-sulfate | 10251-38-4 | 98.0% | 550.75 | 5 MG
SDP
Catalog No. 5000361407
Encompass_Preferred
18β-Glycyrrhetyl-3-O-sulfate is a potent type 2 11β-hydroxysteroid dehydrogenase (11β-HSD2) inhibitor with an IC50 of 0.10 μM using rat kidney microsome. It is the major metabolite of glycyrrhetinic acid (GA) and a substrate of organic anion transporter (OAT) 1 and OAT3. This product exhibits anti-inflammatory effects and shows potential for pseudohyperaldosteronism research.
  • Potent type 2 11β-hydroxysteroid dehydrogenase (11β-HSD2) inhibitor
  • IC50 of 0.10 μM using rat kidney microsome
  • Major metabolite of glycyrrhetinic acid (GA)
  • Substrate of organic anion transporter (OAT) 1 and OAT3
  • Exhibits anti-inflammatory effects
  • Potential for pseudohyperaldosteronism research
Catalog No. 50-003-61407 Supplier Medchemexpress LLC Supplier No. HY1486825MG
18β-Glycyrrhetyl-3-O-sulfate is a potent type 2 11β-hydroxysteroid dehydrogenase (11β-HSD2) inhibitor with an IC50 of 0.10 μM using rat kidney microsome. It is the major metabolite of glycyrrhetinic acid (GA) and a substrate of organic anion transporter (OAT) 1 and OAT3. This product exhibits anti-inflammatory effects and shows potential for pseudohyperaldosteronism research.
  • Potent type 2 11β-hydroxysteroid dehydrogenase (11β-HSD2) inhibitor
  • IC50 of 0.10 μM using rat kidney microsome
  • Major metabolite of glycyrrhetinic acid (GA)
  • Substrate of organic anion transporter (OAT) 1 and OAT3
  • Exhibits anti-inflammatory effects
  • Potential for pseudohyperaldosteronism research