Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 18α-Glycyrrhizic acid | 83896-44-0 | 10 MG
SDP

Catalog No. 5000417904
Encompass_Preferred

18α-Glycyrrhizic acid is a derivative of Glycyrrhizic acid with weak antihepatotoxic and anti-inflammatory activities. This product is for research use only.

  • Purity: 99.79%.
  • Appearance: Solid.
  • Color: White to off-white.
  • Store at -20°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • In vitro solubility: 100 mg/mL in DMSO.
  • Two in vivo dissolution protocols available.

Catalog No. 50-004-17904 Supplier Medchemexpress LLC Supplier No. HYN6911A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

18α-Glycyrrhizic acid is a derivative of Glycyrrhizic acid with weak antihepatotoxic and anti-inflammatory activities. This product is for research use only.

  • Purity: 99.79%.
  • Appearance: Solid.
  • Color: White to off-white.
  • Store at -20°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • In vitro solubility: 100 mg/mL in DMSO.
  • Two in vivo dissolution protocols available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.