Learn More
Bellco Glass, Inc. 18 Hole Large Flange Headplate for Omni Vessels
SDP
Catalog No. 501944900
Encompass_Preferred
200mm SST 18 port headplate for large flange omni vessels. MADE IN THE USA. 12@ 1/4" OD6@ 1/2" OD1 center for drive motorLarge flange headplate assembliesUniversal headplates fit 5l thru 36l vesselsCorrosion resistant 316l stainless steelIncludes red silicone o-ring and 200mm t-bolt clampo-ring replacements:a554-42 6 eacha554-30 12 eacha554-13 1 each
Catalog No. 50-194-4900 Supplier Bellco Glass, Inc. Supplier No. 196488510
Add to Cart
Add to Cart
Request a Quote
200mm SST 18 port headplate for large flange omni vessels. MADE IN THE USA. 12@ 1/4" OD6@ 1/2" OD1 center for drive motorLarge flange headplate assembliesUniversal headplates fit 5l thru 36l vesselsCorrosion resistant 316l stainless steelIncludes red silicone o-ring and 200mm t-bolt clampo-ring replacements:a554-42 6 eacha554-30 12 eacha554-13 1 each