Learn More
Medchemexpress LLC 18-Methoxy-18-oxooct 5g
SDP
Catalog No. 5000204808
Encompass_Preferred
18-Methoxy-18-oxooctadecanoic acid is a long alkane linker with a terminal carboxylic acid group The terminal carboxylic acid group can react with primary amine groups in the presence of activators (e g HATU) to form a stable amide bond
Catalog No. 50-002-04808 Supplier Medchemexpress LLC Supplier No. HYW1057375G
18-Methoxy-18-oxooctadecanoic acid is a long alkane linker with a terminal carboxylic acid group The terminal carboxylic acid group can react with primary amine groups in the presence of activators (e g HATU) to form a stable amide bond