Learn More
Electron Microscopy Sciences 18 Pin Mount Storage Box and Holder
SDP
Catalog No. 50363972
Encompass_Preferred
The EMS Specimen mount Box and holder is a unique storage box for Pin mounts with 3.2 mm (1/8") Pins. The box is numbered 1-18 and it can accept mounts ranging in size from 6.4 mm to 50 mm. With the ability to hold as follows: 18 of 12.7 mm; 8 of 25 mm; 2 of 32 mm and even 38 or 50 mm The maximum specimen height between mount surface and closed lid is 16 mm (5/8").
Catalog No. 50-363-972 Supplier Electron Microscopy Sciences Supplier No. 76506
The EMS Specimen mount Box and holder is a unique storage box for Pin mounts with 3.2 mm (1/8") Pins. The box is numbered 1-18 and it can accept mounts ranging in size from 6.4 mm to 50 mm. With the ability to hold as follows: 18 of 12.7 mm; 8 of 25 mm; 2 of 32 mm and even 38 or 50 mm The maximum specimen height between mount surface and closed lid is 16 mm (5/8").