Learn More
Fisherbrand™ 18 Place Slide Staining Tray
Catalog No. 14127540 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
16.1 x 8.9 x 1.6 in.
Catalog No. Dimensions (L x W x H)
14-127-540 16.1 x 8.9 x 1.6 in.
1 options
Catalog No. 14-127-540
Add to Cart
Add to Cart
Ideal for Clinical and Research laboratory staining
  • Tray holds 18 standard microscope slides (76 x 26 mm) securely in place for staining, rinsing and drying applications
  • Durably constructed from solvent resistant polypropylene blend
  • Non-skid Silicone feet keep tray in place
  • Two ways to empty stain liquid
  • Pour spouts on each corner
  • Drain hole with plug
  • Equipped with handle cut outs for easy handling
  • Black lid included for light sensitive applications
  • Humidity retention wells for specialized staining needs
Specifications
Product Type Slide Staining Tray
Dimensions (L x W x H) 16.1 x 8.9 x 1.6 in.