Learn More
Electron Microscopy Sciences 18 Polygon Spin Bar Assortment Box
SDP
Catalog No. 5027019
Encompass_Preferred
18 Polygon stirring bars: PTFE. consists of 2 each: 10mmx6mm, 15mmx6mm, 20mmx7mm, 25mmx8mm, 30mmx8mm, 40mmx8mm, 50mmx8mm, 60mmx8mm, 80mmx10mm.
Catalog No. 50-270-19 Supplier Electron Microscopy Sciences Supplier No. 6451018
18 Polygon stirring bars: PTFE. consists of 2 each: 10mmx6mm, 15mmx6mm, 20mmx7mm, 25mmx8mm, 30mmx8mm, 40mmx8mm, 50mmx8mm, 60mmx8mm, 80mmx10mm.