Learn More
Electron Microscopy Sciences 18" Polypropylene Retriever
SDP
Catalog No. 5029135
Encompass_Preferred
Polypropylene stirring bar retrievers are ideal for use with all stirring bars. The entire retriever is totally encased in a chemically resistant polypropylene, with a handy ring for hanging when not in use.
Catalog No. 50-291-35 Supplier Electron Microscopy Sciences Supplier No. 6420818
Polypropylene stirring bar retrievers are ideal for use with all stirring bars. The entire retriever is totally encased in a chemically resistant polypropylene, with a handy ring for hanging when not in use.