Learn More
Bio Rad Laboratories 18 WELL 10% SDS CRITERION PREC
Catalog No. NC9830305
Encompass
Criterion Tris-HCl gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 13.3 cm x 8.7 cm x 1.0 mm (W x L x thickness) are supplied in a 15.0 x 10.6 cm cassette and are compatible with Criterio.
Catalog No. NC9830305 Supplier Bio Rad Laboratories Supplier No. 3450010
Criterion Tris-HCl gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 13.3 cm x 8.7 cm x 1.0 mm (W x L x thickness) are supplied in a 15.0 x 10.6 cm cassette and are compatible with Criterio.