Learn More
Euthanex 18 X 15 LID WITH FLOW RESTRICT
Catalog No. NC2319288
Encompass
Euthanex lids allow animals to be gently euthanized through the precise control of gas introduction. The animals remain in their home cages, thereby minimizing added stress. The lid system is highly economical and efficient and still widely used today.Our lids are constructed of heavy-duty, stainless steel. They include an easy, quick disconnect lid stem for gas input as well as a foam gasket for a secure fit.The foam gasket seal allows the lids to be used with smaller cage sizes.Flow restrictors are available and sold separately.
Catalog No. NC2319288 Supplier Euthanex Supplier No. E20045R
Euthanex lids allow animals to be gently euthanized through the precise control of gas introduction. The animals remain in their home cages, thereby minimizing added stress. The lid system is highly economical and efficient and still widely used today.Our lids are constructed of heavy-duty, stainless steel. They include an easy, quick disconnect lid stem for gas input as well as a foam gasket for a secure fit.The foam gasket seal allows the lids to be used with smaller cage sizes.Flow restrictors are available and sold separately.