Learn More
Ace Glass, Inc. 180 x 140mm PTFE support ring with internal bevel. Support ring is useful for supporting columns, flasks, dishes, etc.
SDP
Catalog No. 50977246
Encompass
180 x 140mm PTFE support ring with internal bevel. Support ring is useful for supporting columns, flasks, dishes, etc.
Catalog No. 50-977-246 Supplier Ace Glass, Inc. Supplier No. 1175206
180 x 140mm PTFE support ring with internal bevel. Support ring is useful for supporting columns, flasks, dishes, etc.