Learn More
Alkali Scientific 180L CO2 Incubator H3565-180 W
SDP
Catalog No. 503029556
Encompass_Preferred
Internal microscope option Precision temperature control Excellent accuracy and recovery Heated door and door frame Cell cultures depend on optimized growth conditions in order to thrive With precise temperature and CO control Alkali Scientifics H3565-180 SureTherm180L CO2 Incubator provides the perfect environment for your cultures The Alkali Scientific H3565-180 SureTherm180L CO2 Incubator features a proprietary heat distribution system that combines six sided heating with a low speed internal fan Air is circulated uniformly throughout the chamber without air exchange from the external environment This results in a controlled protected environment with minimal temperature fluctuations (within 0 1C) These are the first and only incubators with our OPTIONAL (patent pending) IncuViewLCI (Live Cell Imaging) technology for viewing cells in real time An internal microscope and large external display allow cells to be viewed without removal from the incubator eliminating the inheren
Catalog No. 50-302-9556 Supplier Alkali Scientific Supplier No. RS3565180
Add to Cart
Add to Cart
Request a Quote
Internal microscope option Precision temperature control Excellent accuracy and recovery Heated door and door frame Cell cultures depend on optimized growth conditions in order to thrive With precise temperature and CO control Alkali Scientifics H3565-180 SureTherm180L CO2 Incubator provides the perfect environment for your cultures The Alkali Scientific H3565-180 SureTherm180L CO2 Incubator features a proprietary heat distribution system that combines six sided heating with a low speed internal fan Air is circulated uniformly throughout the chamber without air exchange from the external environment This results in a controlled protected environment with minimal temperature fluctuations (within 0 1C) These are the first and only incubators with our OPTIONAL (patent pending) IncuViewLCI (Live Cell Imaging) technology for viewing cells in real time An internal microscope and large external display allow cells to be viewed without removal from the incubator eliminating the inheren