Learn More
Alkali Scientific 180L Plus CO2 Incubator H3565-
SDP
Catalog No. 503029678
Encompass_Preferred
These new Alkali ScientificCO2 incubators for mammalian cell culture are loaded with features and options making them among the most versatile and safest cell culture incubators on the market The stainless steel interior delivers excellent temperature uniformity and has seamless rounded corners for quick and easy clean up The digital control panel precisely maintains the six-heating elements that surround the interior for unparalleled uniformity (/- 0 4C) With six-sided heating air-flow is minimized resulting in reduced concern for contamination Dual Infrared Beam sensors track and monitor the CO2 density accurately Transparent sealed inner glass doors allows for visualization of samples without exposing the chamber to ambient temperatures This improves performance and reduces energy waste The inner part of the door including the door frame is heated which helps reduce condensation from forming on the inner glass door The 45L model comes standard with two stainless steel sh
Catalog No. 50-302-9678 Supplier Alkali Scientific Supplier No. RS7518
Add to Cart
Add to Cart
Request a Quote
These new Alkali ScientificCO2 incubators for mammalian cell culture are loaded with features and options making them among the most versatile and safest cell culture incubators on the market The stainless steel interior delivers excellent temperature uniformity and has seamless rounded corners for quick and easy clean up The digital control panel precisely maintains the six-heating elements that surround the interior for unparalleled uniformity (/- 0 4C) With six-sided heating air-flow is minimized resulting in reduced concern for contamination Dual Infrared Beam sensors track and monitor the CO2 density accurately Transparent sealed inner glass doors allows for visualization of samples without exposing the chamber to ambient temperatures This improves performance and reduces energy waste The inner part of the door including the door frame is heated which helps reduce condensation from forming on the inner glass door The 45L model comes standard with two stainless steel sh