Learn More
Chemglass Life Sciences Dish, Crystallizing, 180mL, 80 x 40mm
SDP
Catalog No. 501215016
Encompass_Preferred
Dish, Crystallizing, 180mL, 80 x 40mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.
Catalog No. 50-121-5016 Supplier Chemglass Life Sciences Supplier No. CG8276P80
Dish, Crystallizing, 180mL, 80 x 40mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.