Learn More
Chemglass Life Sciences Dish, Crystallizing, 180mL, 70 x 50mm
SDP
Catalog No. 501215015
Encompass_Preferred
Dish, Crystallizing, 180mL, 70 x 50mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.
Catalog No. 50-121-5015 Supplier Chemglass Life Sciences Supplier No. CG8276P70
Dish, Crystallizing, 180mL, 70 x 50mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.